Notice: The larger size product catalog # C202006 will be discontinued on September 1, 2017. The smaller product (catalog # C202003) will remain available.

One Shot® INVαF´ Chemically Competent E. coli

Catalog number: C202006

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

INVαF´ E. coliallow stable replication of high-copy number plasmids. The genotype of INVαF´ offers the following features:

• Blue/white screening of recombinant clones (lacZΔM15)
• Reduction of homologous recombination of transformed plasmids (recA)
• Increased quality of plasmid DNA preparations (endA1)
• Production and rescue of single-strand DNA from vectors carrying the f1 origin of replication (F´)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Competent Cell Type: Chemically Competent
Species: E. coli
Format: One Shot
Transformation Efficiency: > 1x10^8
Bacterial or Yeast Strain: INVαF´
Product Line: One Shot®
Blue-White Screening: Yes
Cloning Methylated DNA: Yes
Cloning Unstable DNA: Not Suitable for cloning unstable DNA
F' Episome (to make ssDNA): Contains F' Episome
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Improves Plasmid Quality: Yes
Plasmid Type: High Copy Plasmid
Preparing Unmethylated DNA: Not Suitable for preparing unmethylated DNA
Product Size: 40 x 50 µL
Propagating ccdB Vectors: Not for ccdB vector propagation
Reduces Recombination: Yes
T1 Phage - Resistant (tonA): No

Documents

Manuals & protocols