Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

CaptAvidin™ Agarose (Sedimented Bead Suspension)

Catalog number:  C21386

 Related applications: Protein Assays and Analysis | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptAvidin™ biotin-binding protein conjugated to agarose is designed for use in affinity chromatography. CaptAvidin™ biotin-binding protein contains a nitrated tyrosine in its biotin-binding site. This chemical modification permits dissociation of the avidin-biotin complex under milder conditions than the traditional avidin-biotin complex. At pH 4.0, CaptAvidin™ biotin-binding protein binds biotin tightly with a Ka of ~109M-1. This association can then be reversed at pH 10, allowing complete dissociation of the avidin-biotin complex. This is in contrast to the harsh conditions (8 M guanidine hydrochloride, pH 1.5) needed to dissociate the avidin-biotin complex.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Agarose
Product Size: 5 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products