Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

Cascade Blue™ Acetyl Azide, Trisodium Salt

Catalog number:  C2284

 Related applications: Labeling Chemistry | Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The amine-reactive Cascade Blue® acetyl azide and its conjugates yield blue-fluorescence (approximate excitation/emission maxima ~396/410 nm) that can be used as a contrasting color in multicolor applications. When compared with aminocoumarin derivatives, the sulfonated pyrene Cascade Blue® dye shows less spectral overlap with fluorescein. In addition, Cascade Blue® dye exhibits high absorptivity, is highly fluorescent and, unlike most dyes, resists quenching upon protein conjugation.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Cascade Blue®
Chemical Reactivity: Amine
Product Line: CASCADE BLUE®
Product Size: 5 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products