Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells
New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

Calcium Calibration Buffer Kit #1, zero and 10 mM CaEGTA

Catalog number:  C3008MP

 Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Calcium Calibration Buffer Kits are designed to help determine the dissociation constant (Kd) of fluorescent Ca2+ indicators using a fluorometer or quantitative imaging system. The Kd of an indicator can be calculated from a plot generated by scanning the excitation or emission of the indicator in the presence of eleven different Ca2+ concentrations. Calcium Calibration Kit #1 contains 50 mL of 10 mM K2EGTA and 50 mL of 10 mM CaEGTA. Both solutions contain 100 mM KCl and 30 mM MOPS, pH 7.2 and are prepared in deionized water. These stock solutions can be blended to make buffers having free Ca2+ ranging from 0 µM to 39 µM.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Ion Calibration Reagent
For Use With (Equipment): Fluorescence Microscope, Fluorometer
Detection Method: Fluorescent
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products