Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Calcium Orange™, Tetrapotassium Salt, cell impermeant

Catalog number: C3013

Molecular Probes™  Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The cell-impermeant visible light-excitable Ca2+ indicator Calcium Orange™ dye exhibits an increase in fluorescence emission intensity upon binding Ca2+ with little shift in wavelength. Calcium Orange™ dye has an excitation maxium near 550 nm and is compatible with standard tetramethylrhodamine optical filters.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Calcium Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope
Cell Permeability: Cell-Impermeant
Product Size: 500 µg
Detection Method: Fluorescent
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols