Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Calcium Sponge™ S (BAPTA polystyrene)

Catalog number: C3047

Molecular Probes™  Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Calcium Sponge™ S consists of the Ca2+-selective BAPTA chelator coupled to a polymer matrix. Polycations including Ca2+ and Zn2+ can be selective removed from aqueous solutions simply by stirring a solution with the water-insoluble Calcium Sponge S polymer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Chelator (Non-Fluorescent)
Product Size: 1 g
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols