Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

Cascade Blue™ hydrazide, Trilithium Salt

Catalog number:  C3239

 Related applications: Cell Tracing & Tracking | Labeling Chemistry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The cell-impermeant, fixable, polar tracer Cascade Blue® hydrazide can be used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis. This water-soluble tracer can be introduced into cells by whole-cell patch clamping, iontophoresis, osmotic lysis of pinocytic vesicles or comparable methods. The primary aliphatic amine of Cascade Blue® hydrazide can also be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Cascade Blue®
Chemical Reactivity: Aldehyde, Carboxylic Acid, Ketone
Product Line: CASCADE BLUE®
Product Size: 10 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products