Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.
Invitrogen™

Click-IT™ GlcNAz Metabolic Glycoprotein Labeling Reagent (tetraacetylated N-Azidoacetylglucosamine) This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Click-iT® GlcNAz metabolic glycoprotein labeling reagent provides the first part of a simple and robust two-step technique to identify and characterize intracellular O-GlcNAc glycoproteins. In step one, cultured cells are incubated with the azide-modified glucosamine (GlcNAc). The azido-sugar is incorporated into intracellular O-GlcNAc-containing glycoproteins through the permissive nature of the oligosaccharide biosynthesis pathway. In step two, via the chemoselective ligation or click reaction between an azide and an alkyne, the azido-labeled glycoproteins can then detected with a Click-iT® Glycoprotein Detection kit for gels (TAMRA or Dapoxyl® alkyne) or Western blots (biotin alkyne). The Click-iT® glycoprotein products are compatible with LC-MS⁄MS and Multiplexed Proteomics™ technologies for in-depth analyses of the glycoproteome.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Biotin-Based, Fluorescent
Green Features: Less hazardous
Label or Dye: Alexa Fluor™ 488, Alexa Fluor™ 555, Alexa Fluor™ 594, Alexa Fluor™ 647, Biotin, Oregon Green® 488, TMR (Tetramethylrhodamine)
Labeling Target: Proteins (Glycoproteins)
Product Line: Click-iT™
Product Size: 5.2 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products