Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.

Click-IT® Biotin Protein Analysis Detection Kit This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: C33372

Invitrogen™  Related applications: Labeling Chemistry | Protein Assays and Analysis | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Click-iT® Biotin Glycoprotein Detection Kit provides the second part of a simple and robust technique to identify and characterize glycoproteins by Western blot. In step two, after the incorporation of an azide moiety into protein glycan structures with either a Click-iT® metabolic labeling reagent or the Click-iT® Enzymatic Labeling system, the azide-modified glycoproteins are detected via the chemoselective ligation or click reaction between an azide and an alkyne. With this technique, detection sensitivity is in the low femptomole range and biotin-labeled samples can be detected before or after probing the Western with a primary antibody.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Biotin-Based, Fluorescent
Label or Dye: Biotin
Labeling Target: Proteins (Glycoproteins)
Product Line: Click-iT™
Product Size: 1 kit
Green Features: Less hazardous
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products