Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.

Click-iT® O-GlcNAc Peptide LC/MS Standard (H-Thr-Ala-Pro-Thr-(O- GlcNAc)Ser-Thr-Ile-Ala- Pro-Gly-OH) This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: C33374

Molecular Probes™  Related applications: Labeling Chemistry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Click-iT® O-GlcNAc-modified LC⁄MS standard peptide includes the O-GlcNAc modification site found within the primary sequence of the transcription factor, CREB. The peptide is supplied as a standard for MALDI-MS and LC-MS/MS characterization of the O-GlcNAc posttranslational modification.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Mass Range: 1118.5 Da
Product Size: 5 nmol
Calibration Type: LC⁄MS Standards
Mass Standard Type: Peptide Mass Standard
Green Features: Less hazardous
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products