Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Calcium Green™-5N, AM, cell permeant - Special Packaging

Catalog number:  C3739

Invitrogen™  Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. The fluorescence signal from these cells is generally measured using fluorescence microscopy, fluorescence microplate assays, or flow cytometry.

Calcium Indicator (AM Ester) Specifications:
• Label (Ex/Em): Calcium Green™ -5N (506/532 nm)
• Fluorescence intensity increase upon binding Ca2+ : ~38 fold
• Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength


Spectral Characteristics of Molecular Probes® Calcium Indicators
These probes are excited by visible light, and because the energy required for excitation is low, the potential for cellular photodamage is reduced. Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of the spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem.

More Choices for Fluorescent Calcium Indicators
We offer a large selection of Molecular Probes® calcium indicators for use in various experimental scenarios, for example dextran versions for reduced leakage and compartmentalization and BAPTA conjugates for detecting high-amplitude calcium transients. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes® Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes® Handbook.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Calcium Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope
Cell Permeability: Cell-Permeant
Detection Method: Fluorescent
Product Line: Calcium Green™
Product Size: 10 x 50 µg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols