One Shot® BL21(DE3)pLysS Chemically Competent E. coli

Catalog number: C606010

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

BL21(DE3)pLysS cells are ideal for use with T7 promoter-based expression systems (e.g., pRSET, pCR™T7, and pET). BL21(DE3)pLysS E. coli carry both the DE3 lysogen and the plasmid pLysS. pLysS constitutively expresses low levels of T7 lysozyme, which reduces basal expression of recombinant genes by inhibiting basal levels of T7 RNA polymerase.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Competent Cell Type: Chemically Competent
Species: E. coli
Format: One Shot
Blue-White Screening: No
Bacterial or Yeast Strain: BL21(DE3)pLysS, BL21(DE3)
Product Size: 10 x 50 µL
Reduces Recombination: No
Cloning Methylated DNA: No
Improves Plasmid Quality: No
Propagating ccdB Vectors: Not for ccdB vector propagation
Transformation Efficiency: > 1x10^8
F' Episome (to make ssDNA): Lacks F' Episome
T1 Phage - Resistant (tonA): No
High Throughput Compatibility: Not High Throughput-Compatible (Manual)

Documents

Manuals & protocols

Recommended products