Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

6-ROX, SE (6-Carboxy-X-Rhodamine, Succinimidyl Ester), single isomer

Numéro de catalogue:  C6126

Invitrogen™  Applications liées: Labeling Chemistry | Protein Labeling & Crosslinking

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

The isomeric 6-ROX, SE is an amine-reactive form of carboxy-X-rhodamine and is widely used for oligonucleotide labeling and automated DNA sequencingapplications.
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Label or Dye: ROX™
Chemical Reactivity: Amine
Product Size: 5 mg
Shipping Condition: Room Temperature

Contenu et stockage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuels et protocoles

Produits recommandés