Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

5(6)-CR 6G, SE (5-(and-6)-Carboxyrhodamine 6G, Succinimidyl Ester), mixed isomers

Numéro de catalogue:  C6157

Invitrogen™  Applications liées: Labeling Chemistry | Protein Labeling & Crosslinking

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

The excitation and emission spectra of carboxyrhodamine 6G (CR 6G) fall between those of fluorescein and tetramethylrhodamine. With a peak absorption at 525 nm, conjugates of carboxyrhodamine 6G are an excellent match to the 514 nm spectral line of the argon-ion laser. The amine-reactive 5(6)CR 6G, SE can be used to attach this fluorophore to biomolecules of interest.
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Label or Dye: Rhodamine 6G
Chemical Reactivity: Amine
Product Size: 5 mg
Shipping Condition: Room Temperature

Contenu et stockage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuels et protocoles

Produits recommandés