CBQCA Protein Quantitation Kit

Catalog number: C6667

Molecular Probes™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The CBQCA Protein Quantitation Kit is a very sensitive assay for quantitating proteins in solution, capable of detection as low as 10 ng of protein per mL. Similar in sensitivity to our NanoOrange protein quantitation reagent (N-6666), CBQCA is better suited for accurate quantitation of proteins in the presence of lipids, membrane fractions or detergents, and for lipoproteins and small peptides.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: Protein Quantitation Assay
Detection Method: Fluorescent
For Use With (Equipment): Microplate Reader
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products