One Shot® INV110 Chemically Competent E. coli

Catalog number: C717103

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

INV110 E. coli are derived from JM110 E. coli and are specifically designed for the growth and purification of plasmid DNA that will be digested with dam or dcm methylation-sensitive restriction enzymes. INV110 is a significant improvement over the JM110 strain. INV110 has been engineered to carry an endA1 mutation, which eliminates the non-specific endonuclease Endonuclease I, for improved plasmid DNA preparation. In addition, the INV110 genotype features:

• Blue/white screening of recombinant clones (lacZΔM15)
• Reduced cleavage of foreign methylated DNA for improved transformation of genomic DNA [Δ(mcrC-mrr)]
• F´ episome for phage infection and rescue of single-strand DNA
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Competent Cell Type: Chemically Competent
Species: E. coli
Format: One Shot
Transformation Efficiency: > 1x10^6
Bacterial or Yeast Strain: INV110
Product Line: One Shot®
Blue-White Screening: Yes
Cloning Methylated DNA: No
Cloning Unstable DNA: Not Suitable for cloning unstable DNA
F' Episome (to make ssDNA): Contains F' Episome
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Improves Plasmid Quality: Yes
Preparing Unmethylated DNA: Suitable for preparing unmethylated DNA
Product Size: 20 x 50 µL
Propagating ccdB Vectors: Not for ccdB vector propagation
Reduces Recombination: No
T1 Phage - Resistant (tonA): Yes

Contents & storage

One Shot® INV110 Chemically Competent E. coli contains 10 transformations of chemically competent cells in One Shot® format. Store at -80°C.

Documents

Manuals & protocols

Recommended products