Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

ChromaTide™ Texas Red™-12-dUTP

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Molecular Probes® ChromaTide® dye labeled dUTP, OBEA-dCTP, and UTP nucleotides can be used to synthesize labeled DNA probes without the need for hazardous and expensive radioisotope-labeled nucleotides. These nucleotides can be incorporated using standard molecular biology techniques; labeled probes can then be used in in situ hybridization, microarray, or blotting protocols. ChromaTide® dye labeled nucleotides are available in different fluorescent colors to facilitate multicolor analysis.

ChromaTide® Labeled Nucleotides Specifications:
• Ex/Em of dye: Texas Red®-12-dUTP (595/615 nm)
• Length of alkynylamino linker: 12 atoms


Methods for Incorporating ChromaTide® Nucleotides Into Probes
• Nick translation
• Random primer labeling
• End-labeling with terminal deoxynucleotidyl transferase
• Reverse transcription
• PCR amplification


See Methods for Enzymatic Incorporation of ChromaTide® dUTPs for specific guidelines for each of these methods.

Alexa Fluor® and BODIPY® Fluorescent Dyes Make Excellent Probes
Probes made with labeled nucleotides can be used for multicolor techniques such as in situ hybridization and hybridization to arrays. Our proprietary BODIPY® and Alexa Fluor® dye conjugates are exceptionally bright, photostable, and essentially pH insensitive. The narrow emission profile of the BODIPY® dyes helps ensure minimal spectral overlap. The Alexa Fluor® dyes are highly water soluble, as are DNA probes made from them, making them the labels of choice for fluorescence in situ hybridization.

Long Linkers Improve Performance
The ChromaTide® dUTP and UTP nucleotides are modified at the C-5 position of uridine via a unique alkynylamino linker, which provides a spacer between the nucleotide and the dye to reduce interactions between them. The number in the product name, e.g., the “12” in fluorescein-12-dUTP, indicates the net length of the spacer, in atoms. These spacers result in brighter conjugates and increased hapten accessibility for secondary detection reagents.

For complete listing of our ChromaTide® Reagents: Molecular Probes ChromaTide® and aha labeled nucleotides—Table 8.5.
For additional information on these labeling reagents, read Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes® Handbook.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration: 1 mM
Label or Dye: Texas Red®
Labeling Method: Direct Labeling
Product Line: ChromaTide®, Texas Red®
Product Size: 25 µL
Shipping Condition: Wet Ice

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols