MultiShot™ StripWell Mach1™ T1 Phage-Resistant Chemically Competent E. coli

Catalog number: C869601

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

MultiShot™ StripWell Mach1™ T1 Phage-Resistant Chemically Competent E. coli are the fastest growing chemically competent cells currently available (see figure). Doubling time is approximately 50 minutes, compared to >74 minutes for other cloning strains. Mach1™ colonies are clearly visible within eight hours of plating the transformation mix, enabling you to plate and pick colonies the same day. Perform mini-preps after only 4 hours of growth from an overnight colony—saving an entire day in your experiments. The fast-growing Mach1™ T1R strain:

• Minimizes nonspecific recombination in cloned DNA (recA)
• Offers resistance to T1 and T5 phage (tonA)
• Yields cleaner preparations of DNA and better results in downstream applications due to the elimination of nonspecific digestion by Endonuclease I (endA1)
• Provides efficient transformation of unmethylated DNA from PCR amplifications (hsdR)
• Allows blue/white screening of recombinant clones (lacZΔM15)

Two formats to choose from
• The convenient One Shot® format provides single-use, 50-µL aliquots that eliminate efficiency-zapping, freeze-thaw cycles and money wasted on unused cells
• The MultiShot™ StripWell format allows rapid, high-throughput transformation in a 96-well stripwell plate, which allows you to perform as few or as many transformations as you need without wasting reagents
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Competent Cell Type: Chemically Competent
Species: E. coli
Format: MultiShot StripWell
Transformation Efficiency: > 1x10^9
Bacterial or Yeast Strain: Mach1™
Fast Growth: Yes
Product Size: 1 plate
Blue-White Screening: Yes
Cloning Unstable DNA: Not Suitable for cloning unstable DNA
Reduces Recombination: Yes
Cloning Methylated DNA: No
Improves Plasmid Quality: Yes
Propagating ccdB Vectors: Not for ccdB vector propagation
F' Episome (to make ssDNA): Lacks F' Episome
Preparing Unmethylated DNA: Not Suitable for preparing unmethylated DNA
T1 Phage - Resistant (tonA): Yes
High Throughput Compatibility: High Throughput-Compatible

Contents & storage

Contains:
• Mach1™ T1 Phage-Resistant (T1R) Chemically Competent E. coli: One 96-stripwell plate, 50 µl per well
• pUC19 DNA (10 pg/µl): 1 vials, 50 µl
• S.O.C. medium: 2 bottles, 15 ml each

Store Competent Cells at -80°C. Store pUC19 DNA at -20°C. Store SOC Medium at 4°C or at room temperature.

Documents

Manuals & protocols

Recommended products