c-Met (Soluble) Human Matched Antibody Pair

Catalog number: CHO0315

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This antibody pair Kit comes with matched antibody pairs to detect and quantify protein levels of soluble c-Met. Activation of c-Met, also known as HGF (hepatocyte growth factor receptor), regulates RAS signalling and cell scattering. Also available is Buffer Kit for Antibody Pairs that saves time with premade, easy-to-use buffers and solutions optimized for use with Invitrogen Antibody Pairs. The Assay Buffer can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate. Buffer reagents needed to complete the reaction are sold separately under Catalog #CNB0011.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene Symbol: MET
Assay Sensitivity: See product documentation
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Gene Aliases: AUTS9, DFNB97, HGFR, RCCP2, c-Met
Gene ID: 4233
Gene Name: MET proto-oncogene, receptor tyrosine kinase
Immunoassay Kit Format: Matched Antibody Pair
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 5 x 96 tests
Protein Name: Hepatocyte growth factor receptor
Protein Subtype: Receptor Tyrosine Kinases
Shipping Condition: Wet Ice
Target (Kit Named): c-Met
Target Molecule: Protein
Target Species (Named): Human
Target Species (Validated): Human
Target Sub-Specificity: Soluble
UniProt ID: P08581

Contents & storage

Antibody pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet.
Store at 2-8°C.

Documents

Manuals & protocols

Recommended products