Cocoa Sample Recovery (CSR) Broth
Thermo Scientific™  Applications liées: Industrial Microbiology
Test for Salmonella species in chocolate products with ease and reduce time to results using Thermo Scientific™ Cocoa Sample Recovery (CSR) Broth. This novel, single-step enrichment medium is designed to recover and grow low levels of stressed Salmonella from raw materials and interim products, such as cocoa powder, liquor, mass and butter, as well as finished chocolate products.
Error lors du chargement de votre contenu!
  Numéro de catalogue Prix ({{currency}}) Disponibilité Qté
{{product.sku}}
également connu comme {{product.formattedSku}} 
{{product.availability.message}} Ajouter au panier
{{addItemToList.successMessage}} {{addItemToList.listName}}
Introducing shared lists
Haut de la page
Description
The innovative formulation of CSR Broth not only enables faster recovery and growth of Salmonella, but also neutralizes the PCR-inhibiting properties commonly associated with these types of samples.
  • Fast results: optimized formulation enables enrichment in 20 hours, compared to traditional methods that can take up to 39 hours, reducing time to result and streamlining your workflow
  • Proven performance: tested on some of the most challenging cocoa-containing ingredients and finished chocolate products, this novel formulation reduces the bactericidal effects of polyphenols often found in cocoa and chocolate products, providing an optimized environment for recovery and growth of microorganisms
  • Versatility: maximize your detection by using before culture, immunological or PCR methods for maximum efficiency and detection
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Documents
Manuels et protocoles