TNF-alpha Mouse Antibody Pair

Catalog number: CMC3013

Novex™  Related applications: Cell Viability, Proliferation & Function | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Antibody Pair Kit comes with matched antibody pairs to detect and quantify protein levels of mouse TNF-α (Tumor Necrosis Factor-α). Also available is Buffer Kit for Antibody Pairs that saves time with premade, easy-to-use buffers and solutions optimized for use with Invitrogen Antibody Pairs. The Assay Buffer can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate. Buffer reagents needed to complete the reaction are sold separately under Catalog No. CNB0011.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Accession Number: NM_013693.2, NP_038721.1
Emission Class: Visible
For Use With (Equipment): Microplate Reader
Form: Concentrated
Gene Aliases: DIF, MGC151434, TNF-alpha, TNFSF2, TNFalpha, Tnfa, Tnfsf1a
Gene ID: 21926
Gene Symbol: TNF
Label or Dye: HRP (Horseradish Peroxidase)
Marker: TNF-α
Product Size: 10 plates
Protein Family: Cytokines & Receptors
Quantity: 10×96-well plates
Reactivity: Mouse
Research Category: Apoptosis, Immunology
Target Gene: TNF-α
Target Molecule: Protein
Validated Application: ELISA
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Antibody pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet.
Store at 2-8°C.

Documents

Manuals & protocols

Recommended products