ChargeSwitch® NoSpin Plasmid Micro Kit This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: CS1020110

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Sample size 0.5-1 ml overnight culture; Typical yield up to 5 µg

• Provides centrifuge-free purification, ideal for automation
• Avoids introduction of enzymatic contaminants for better results
• Optimized for automation

The ChargeSwitch® NoSpin Kits provide centrifuge-free microprep plasmid purification, ideal for automation. The novel NoSpin reagent causes bacterial cell flocculation and allows cells to be pelleted without the need for automation bottlenecks such as centrifugation. This kit includes all the necessary reagents for the number of preps listed and requires the use of a magnetic accessory. Please see the Accessories Chapter (Chapter 6) to select the best magnetic accessory for your needs.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

High Throughput Compatibility: High Throughput-Compatible
Sample Type (General): Bacteria
Downstream Application: Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Final Product: Plasmid DNA
Isolation Technology: ChargeSwitch® Chemistry
Product Line: ChargeSwitch®
Product Size: 960 preps
Sample Type (Specific): Bacterial Culture
System: ChargeSwitch®
Green Features: Fewer resources used, Less hazardous, Less waste
Shipping Condition: Room Temperature

Contents & storage

All components of the ChargeSwitch® NoSpin Plasmid Kits are shipped at room temperature. Upon receipt, store as follows: Store ChargeSwitch® Precipitation Buffer (N5) at 4“C. Mix the RNase I in ChargeSwitch® Resuspension Buffer (R4) and store the resulting solution at 4“C. Store all other components at room temperature. All components are guaranteed stable for 6 months if stored properly.

Components:

ChargeSwitch® MagnaClear™ Beads
ChargeSwitch® Magnetic Beads
ChargeSwitch® Resuspension Buffer (R4; 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
ChargeSwitch® Lysis Buffer (L9)
ChargeSwitch® Precipitation Buffer (N5)
ChargeSwitch® Wash Buffer (W11)
ChargeSwitch® Wash Buffer (W12)
ChargeSwitch® Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
RNase A (5 mg/ml in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)

Documents

Manuals & protocols