ChargeSwitch® Direct gDNA Purification Kit, 96-well

Catalog number: CS11205

Invitrogen™  Related applications: Automated Nucleic Acid Purification | DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ChargeSwitch® Direct 96 gDNA Kit is a solid-phase, high-throughput genomic DNA (gDNA) purification system designed for rapid and efficient isolation of gDNA from fresh or frozen samples. This kit utilizes 96-well plates coated with the ChargeSwitch® nucleic acid-binding surface, whose ionic charge can be switched by changing the pH of the surrounding buffer. At low pH, the positive charge of the ChargeSwitch® surface binds the negatively charged nucleic acids. Proteins and other contaminants that do not bind are washed away, leaving pure DNA ready for downstream applications. This kit is designed for research use only (Limited Use Label License 5).


– Successful yields from very small samples volumes--up to 10 µl blood or 2 x 104 cells
– Highly pure DNA isolated using gentle buffers--no guanidine or ethanol
– No sample transfer that can lead to error or contamination--direct amplification in the same well as DNA binding without elution
– Adaptable to your throughput needs--compatible with most automated liquid handling platforms



Contents and Storage

The ChargeSwitch® Direct 96 gDNA Kit contains ChargeSwitch® Plate Binding and Lysis Buffer, ChargeSwitch® Wash Buffers, ChargeSwitch® Elution Buffer, ChargeSwitch® Direct 96 gDNA Plate(s), and Plate Seal(s). Store all components at room temperature. The plates can be stored for up to 12 months.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Blood, Cells
Isolation Technology: ChargeSwitch® Chemistry
Downstream Application: PCR, Cloning, Southern Blotting, Sequencing
High Throughput Compatibility: High Throughput-Compatible
Product Size: 96 preps
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols