Chargeswitch™ Pro Filter Plasmid Maxi Kit

Catalog number: CS31107

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Chargeswitch® -Pro Filter Plasmid Maxi plasmid kit includes 25 plasmid maxipreps and provides high yield and high purity plasmid with the fastest and the simplest protocol.
The ChargeSwitch®-Pro Plasmid Filter Mini, Midi, Maxi Kits generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications. The system is based upon a novel dual⁄nested column design which facilitate the fastest and simplest protocol for high quality plasmid. The system is composed of two columns. The inner flat bottomed column, also referred to as “lysate clarification" column, contains filter materials suitable for rapid clarification of bacterial lysate. The outer luer bottomed column, also referred to as “binding" column, contains ChargeSwitch™ (CST) derivatized membrane that immediately binds plasmid DNA from the clarified lysate. This nested-column design enables simultaneous clarification and DNA binding in a single short centrifugation (or vacuum) step. The combined advantages of novel, optimized plastics and a unique chemistry are an extreme competitive advantage of speed and convenience.



To accomplish this, the ChargeSwitch®-Pro Filter Kit provides:

• 100% ethanol-free, guanidinium-free, organics-free, aqueous protocol for a safer, more streamlined method
• Cleaner purified product for improved performance in downstream applications
• A novel Dual⁄nested column design for simultaneous lysate clearing and binding step which offers the fastest and the simplest protocol
• Endotoxin units < 10 EU ⁄ ug DNA for
• Vacuum Elution can be done with the EveryPrep™ Universal Vacuum Manifold (SKU # K211101)



How it works
The ChargeSwitch® technology replaces the use of ethanol, chaotropic salts, or organic solvents for nucleic acid purification with a simple ion-exchange mechanism. At low pH values (<6.5) the ChargeSwitch® surface ligand is positively charged and binds plasmid DNA. Raising the pH (>8.5) neutralizes the charge and elutes the bound nucleic acid. The ChargeSwitch® protocol retains the ease and familiarity of the common silica spin column protocol with a binding, washing, and elution step, making adapting to the ChargeSwitch® method effortless.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Sample Type (General): Bacteria
Downstream Application: Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Elution Volume - max: 2 ml
Elution Volume - min: 1 ml
Final Product: Plasmid DNA
Format: Spin Column
Isolation Technology: ChargeSwitch® Chemistry
Product Line: ChargeSwitch®
Product Size: 25 preps
Purification Time: 30 min
Purity or Quality Grade: Transfection Grade
Quantity: 25 columns
Sample Type (Specific): Bacterial Culture
Starting Material (Volume): 100 ml
System: ChargeSwitch®
Yield (up to): 800 µg
Shipping Condition: Room Temperature

Contents & storage

The ChargeSwitch®-Pro Plasmid Kit contains Columns with Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffers 1 and 2, and Elution Buffer. Store at room temperature. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products