Gibco™

IL6 (Interleukin 6) CTS™ Recombinant Human Protein

Catalog number:  CTP0061

 Related applications: Clinical & Translational Research | Mammalian Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Cell Therapy Systems (CTS™) Recombinant Human Interleukin 6 (IL6) is a bioactive protein intended for use in cell culture applications, including pre-clinical and clinical cell therapy studies. IL6 is a pleotropic cytokine involved in several cellular processes including inflammation, hematopoeisis, and immune response. The lyophilized, E. coli-expressed IL6 CTS™ Recombinant Human is verified for activity through dose-dependent proliferation of mouse B9 cells.

Qualified—helps reduce your burden in qualifying reagents for regulatory submission in cell therapy applications
Highly Pure—minimizes interference from other proteins or contaminants
Active—get more results with less protein

Simplified Transition into Clinical Research
GIBCO® Cell Therapy Systems (CTS™) products are of high quality and supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The GIBCO® CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

High Purity
To help verify that GIBCO® CTS™ Recombinant growth factors are of the highest quality, each protein is analyzed for purity, along with structural homogeneity, to help ensure a biologically active protein. The purity of each GIBCO® CTS™ recombinant growth factor is verified by SDS-PAGE to be over 95% pure and to have endotoxin levels below 0.1 ng/μg.

High Biological Activity
All GIBCO® CTS™ Recombinant growth factors are verified for activity. Each lot has its own ED50 determined using an activity assay based on a specific function of the growth factor. The activity of our IL6 CTS™ Recombinant Human growth factor is determined by the dose-dependent proliferation of mouse B9 cells to have an ED50 range of 0.008–0.02 ng/mL (specific activity: 1.25 × 10^8–5.0 × 10^7 units/mg). Optimal concentration for individual applications should be determined by a dose response assay.

Related Links
Learn More About GIBCO® Cell Therapy Systems (CTS™) Products
Cell Therapy Systems Technical Support Web Page
Our Full Collection of GIBCO® CTS™ Growth Factors
Our Full Collection of GIBCO® Growth Factors
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Bone Research, Clinical Research, Immunology, Inflammation, Stem Cell Research
Activity: 0.008 ng/ml
Bioassay: Proliferation of B9 cells
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Gene Aliases: IL-6 (Interleukin 6)
Molecular Weight: 21.3 kDa
Product Line: CTS™
Product Size: 100 µg
Protein Family: Cytokines & Receptors
Protein Form: Full Length, Ligand, Recombinant
Protein Length or Portion (a.a.): 184 aa
Protein Subtype: Interleukins
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

2° to 8° C

Documents

Manuals & protocols

Recommended products