IL7 (Interleukin 7) CTS™ Recombinant Human Protein

Catalog number: CTP0073

Gibco™  Related applications: Clinical & Translational Research | Mammalian Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant IL7 is a bioactive protein intended for use in cell culture applications. IL7 stimulates the proliferation of pre-B and pro-B cells. IL7 also supports the maturation of megakaryocytes. ED50 range = 0.2 to 1 ng⁄mL (Specific Activity: 5.0 x 10^6 to 1.0 x 10^6 units⁄mg), determined by the dose dependent proliferation of human PBMCs. Optimal concentration for individual application should be determined by a dose response assay.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Specifications

Research Category: Clinical Research, Drug Development, Immunology, Oncology, Signal Transduction, Stem Cell Research
Activity: 0.807 ng/ml
Bioassay: Proliferation of human PBMCs
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Gene Aliases: IL-7 (Interleukin 7)
Molecular Weight: 17.5 kDa
Product Line: CTS™, Gibco®
Product Size: 1 mg
Protein Family: Cytokines & Receptors
Protein Form: Full Length, Ligand, Recombinant
Protein Subtype: Interleukins
Purification: Gel-Purified
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Reactivity: Human
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Lyophilized protein can be stored at 2-8°C dessicated. Once reconstituted, store in aliquots at ≤20°C. Avoid multiple freeze⁄thaw cycles.

Documents

Manuals & protocols

Recommended products