GM-CSF CTS™ Recombinant Human Protein

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications. Recombinant human GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils. ED50 range = 0.02 to 0.1 ng⁄mL, determined by the dose dependent proliferation of human TF-1 cells. Optimal concentration for individual application should be determined by a dose response assay.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Purity: 95 %
Species: Human
Activity: 0.055 ng/ml
Bioassay: Proliferation of TF1 cells
Expression: E. coli
Reactivity: Human
Gene Aliases: GM-CSF
Product Size: 100 µg
Protein Form: Full Length, Recombinant, Ligand
Purification: Gel-Purified
Protein Family: Cytokines & Receptors
Endotoxin Level: < 0.1 ng/µg
Protein Subtype: GM-CSF, G-CSF, & M-CSF
Molecular Weight: 14 kDa
Research Category: Signal Transduction, Differentiation, Stem Cell Research, Clinical Research, Oncology, Drug Development
Validated Application: Cell Culture
Purity or Quality Grade: Carrier-Free
Shipping Condition: Wet Ice

Contents & storage

Lyophilized protein can be stored at 2-8°C dessicated. Once reconstituted, store in aliquots at ≤20°C. Avoid multiple freeze⁄thaw cycles.

Documents

Manuals & protocols

Recommended products