TNF CTS™ Recombinant Human Protein

Catalog number: CTP3013

Gibco™  Related applications: Mammalian Cell Culture | Primary Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant TNF is a bioactive protein intended for use in cell culture applications. TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis. ED50 range = 0.02 to 0.05 ng⁄mL (Specific Activity: 2 x 10^7 to 5 x 10^7 units⁄mg), determined by the dose dependent cytotoxic effect on L929 cells in the presence of actinomycin D.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Purity: 95 %
Species: Human
Activity: 0.034 ng/ml
Bioassay: Cytotoxicity of L929 cells
Expression: E. coli
Reactivity: Human
Gene Aliases: TNF-α
Product Size: 1 mg
Protein Form: Recombinant, Ligand
Purification: Gel-Purified
Protein Family: Cytokines & Receptors
Endotoxin Level: < 0.1 ng/µg
Protein Subtype: TNF (Tumor Necrosis Factor)
Molecular Weight: 17.5 kDa
Research Category: Immunology, Inflammation, Stem Cell Research, Clinical Research, Oncology, Signal Transduction
Validated Application: Cell Culture
Purity or Quality Grade: Carrier-Free
Shipping Condition: Wet Ice

Contents & storage

Lyophilized protein can be stored at 2-8°C dessicated. Once reconstituted, store in aliquots at ≤20°C. Avoid multiple freeze⁄thaw cycles.

Documents

Manuals & protocols

Recommended products