Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Deoxyribonuclease I, Alexa Fluor™ 594 Conjugate

Catalog number: D12372

Invitrogen™  Related applications: Cell Structure

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The bright red fluorescent Alexa Fluor® 594 DNase I selectively binds to G-actin (globular) and can be used for the detection of unpolymerized actin in fixed cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Alexa Fluor™ 594
Sub-Cellular Localization: Actin, Cytoskeleton
Excitation⁄Emission (nm): 590⁄617
Product Line: Alexa Fluor™
Product Size: 5 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products