7-Diethylaminocoumarin-3-Carboxylic Acid

Catalog number: D1421

Molecular Probes™  Related applications: Labeling Chemistry | Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The amine-reactive coumarin, 7-diethylaminocoumarin-3-carboxylic acid can be used to create blue-fluorescent bioconjugates following carbodiimide-activation of the carboxylic acid on the dye and reaction with amines. When compared with AMCA conjugates, conjugates of the UV-light-excitable 7-dialkylaminocoumarin fluorophore have slightly longer-wavelength emission spectra (~470 nm).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Methylcoumarin
Chemical Reactivity: Amine
Product Size: 100 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products