Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

5-DTAF (5-(4,6-Dichlorotriazinyl) Aminofluorescein), single isomer

Numéro de catalogue:  D16

Invitrogen™  Applications liées: Labeling Chemistry | Protein Labeling & Crosslinking

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

5-(4,6-dichlorotriazinyl)aminofluorescein (5-DTAF) is a reactive dye with absorption/emission maxima of ~492/516 nm. Dichlorotriazines readily modify amines in proteins, and are among the few reactive groups that are reported to react directly with polysaccharides and other alcohols in aqueous solution, provided that the pH is >9 and that other nucleophiles are absent.
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Label or Dye: FITC (Fluorescein)
Chemical Reactivity: Alcohol, Amine
Product Size: 100 mg
Shipping Condition: Room Temperature

Contenu et stockage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuels et protocoles

Produits recommandés