Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Diaminobenzidine (DAB) Histochemistry Kit #3, with Streptavidin—HRP

Catalog number: D22187

Molecular Probes™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The DAB Histochemistry Kit for biotinylated probes contains sufficient materials to stain approximately 200 slides using horseradish peroxidase (HRP) and the HRP substrate DAB. The brown-colored DAB reaction product can be visualized by bright-fieldd light microscopy or, following osmication, by electron microscopy.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols