Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Dabcyl, SE, 4-((4-(dimethylamino)phenyl)azo)benzoic Acid, Succinimidyl Ester

Catalog number: D2245

Invitrogen™  Related applications: Labeling Chemistry | Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The amine-reactive quencher, dabcyl succinimidyl ester has a broad and intense visible absorption but no fluorescence making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Dabcyl
Chemical Reactivity: Amine
Product Size: 100 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products