Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

4-Di-1-ASP (4-(4-(Dimethylamino)styryl) -N-Methylpyridinium Iodide)

Catalog number:  D288

Invitrogen™  Related applications: Cell Structure | Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

DASPMI (4-Di-1-ASP) is a styryl dye stains mitochondria of live cells. This dye has a large fluorescence Stokes shift and is taken up relatively slowly as a function of membrane potential.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Orange
For Use With (Equipment): Fluorescence Microscope
Sub-Cellular Localization: Mitochondria
Detection Method: Fluorescent
Product Size: 1 g
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products