Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

4-Di-10-ASP (4-(4-(Didecylamino)styryl)-N-Methylpyridinium Iodide)

Catalog number:  D291

Invitrogen™  Related applications: Cell Tracing & Tracking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The lipophilic aminostyryl probe 4-Di-10-ASP contains a C10 alkyl chain and inserts into membranes with its two alkyl tails and its fluorophore orientted parallel to the phospholipid acyl chain. When this dialkylaminostyryl probe binds to membranes, it exhibits a strong fluorescence enhancement; its fluorescence in water is minimal.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Yellow
For Use With (Equipment): Fluorescence Microscope
Sub-Cellular Localization: Cell Membranes & Lipids
Detection Method: Fluorescent
Product Size: 25 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products