Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

OxyBURST® Green H2DCFDA, SE

Catalog number: D2935

Molecular Probes™  Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

OxyBURST® Green H2DCFDA succinimidyl ester (2',7'-dichlorodihydrofluorescein diacetate, SE) is an amine-reactive assay reagent that can be used to prepare oxidation-sensitive conjugates of a wide variety of biomolecules and particles, including antibodies, antigens, peptides, proteins, dextrans, bacteria, yeast and polystyrene microspheres. Following conjugation to amines, the two acetates of OxyBURST® Green H2DCFDA can be removed by treatment with hydroxylamine at neutral pH to yield the dihydrofluorescein conjugate. OxyBURST® Green H2DCFDA conjugates are nonfluorescent until they are oxidized to the corresponding fluorescein derivatives.

ROS Indicator Specifications:
• Ex/Em: ~492–495/517–527 nm
• Product is air sensitive and should be stored under dry argon or nitrogen
• Product may be dissolved in DMSO, DMF, or ethanol for use
• Indicator is cell permeant (cell loading protocols are available in the literature)
• Fluorescence can be monitored using a flow cytometer, fluorometer, microplate reader, or fluorescence microscope, using excitation sources and filters appropriate for fluorescein


Find More ROS Indicators
We offer an assortment of Molecular Probes® products for the generation of reactive oxygen species (ROS), including singlet oxygen, superoxide, hydroxyl radical and various peroxide and hydroperoxides, as well as for their fluorometric detection in solution. Review Generating and Detecting Reactive Oxygen Species—Section 18.2 in the Molecular Probes® Handbook for more information on these products.

For Research Use. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Free Radical Sensor
Indicator Properties: Fluorescent Dye-Based
Product Size: 5 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products