Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
BODIPY® TR Ceramide
Catalog number: D7540
Invitrogen™  Related applications: Cell Structure
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
BODIPY TR ceramide is a red-fluorescent dye that can be used to stain Golgi apparatus in live cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line: BODIPY®
Product Size: 250 µg
Reagent Type: Sphingolipids
Shipping Condition: Room Temperature
Contents & storage
Store in freezer (-5 to -30°C) and protect from light.
Documents
Manuals & protocols
Recommended products