Want sharper images? Try new Prolong™ Glass Antifade Mountant with refrative index of 1.52 (after curing).

DiD' solid; DiIC18(5) solid (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindodicarbocyanine, 4-Chlorobenzenesulfonate Salt)

Catalog number:  D7757

Invitrogen™  Related applications: Cell Tracing & Tracking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The far-red fluorescent, lipophilic carbocyanine DiD is a longer-wavelength DiI analog. It is supplied as a solid form to facilitate direct application of crystals to membranes and is weakly fluorescent in water but highly fluorescent and quite photostable when incorporated into membranes. It has an extremely high extinction coefficient and short excited-state lifetimes (~1 nanosecond) in lipid environments. Once applied to cells, the dye diffuses laterally within the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Red
For Use With (Equipment): Fluorescence Microscope
Sub-Cellular Localization: Cell Membranes & Lipids
Detection Method: Fluorescent
Product Size: 10 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products