Gene Pool™ cDNA, from human adult normal thyroid tissue

Catalog number: D812501

Invitrogen™  Related applications: cDNA Libraries & Library Construction

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

In addition to the previously listed Gene Pool™ cDNA, the following selection of cDNA from adult human normal, tumor, or disease tissues as well as human fetal normal tissue is available. See Gene Pool™ cDNA for description and storage information. All Special Order Products are subject to tissue/tumor availability. Please call for information. Allow up to two weeks for delivery.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Age: Adult
Tissue: Thyroid
Species: Human
Product Size: 20 reactions
Sample Source: Normal Tissue
Shipping Condition: Dry Ice

Contents & storage

Store in ultra-cold freezer (-68 to -85°C).

Documents

Manuals & protocols