Dog (Beagle) S9 Fractions

Catalog number:  DGS9PL

Gibco™  Related applications: Hepatocytes & Hepatic Models | Pharma & Biopharma | Target-Based ADME⁄Tox Assays

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Liver S9 fractions prepared from a pool of male Beagle Dogs.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Donor Source: Pooled
Format: Tube(s)
Gender: Male
Sub-Cellular Localization: S9 Fraction
Tissue: Liver
Concentration: 20 mg⁄ml
Form: Frozen
Product Line: Gibco®
Product Size: 1 mL
Purity or Quality Grade: Research Grade
Shipping Condition: Dry Ice
Species: Dog⁄Canine

Contents & storage

Store between -60°C and -85°C.

Documents

Manuals & protocols

Recommended products