Want to detect cells in hypoxic condition? Try new fixable Image-iT™ Hypoxia reagent, compatible with most fluorescent platforms.
Invitrogen™

Escherichia coli BioParticles™ Opsonizing Reagent

Catalog number:  E2870

 Related applications: Cell Structure | Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Escherichia coli BioParticles® opsonizing agent is derived from highly purified rabbit polyclonal IgG antibodies and specifically designed to enhance the uptake of the Escherichia coli BioParticals® conjugates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Fluorescence Microscope
Label or Dye: Unconjugated
Product Line: BioParticles™
Product Size: 1 unit
Species: E. coli
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products