Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

EnzChek™ Myeloperoxidase (MPO) Activity Assay Kit

Catalog number:  E33856

 Related applications: Cell Viability, Proliferation & Function | Cellular Imaging | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Myeloperoxidase (MPO) is a unique peroxidase that, in addition to its peroxidation activity, also catalyzes the conversion of hydrogen peroxide (H2O2) and chloride (Cl-) to hypochlorous acid (HOCl). The EnzChek® Myeloperoxidase (MPO) Activity Assay Kit provides assays for the determination of both chlorination and peroxidation activities of MPO in solution and in cell lysates. For detection of chlorination, the kit includes nonfluorescent 3'-(p-aminophenyl) fluorescein (APF), which is selectively cleaved by hypochlorite (-OCl) to yield fluorescein. Peroxidation is detected using nonfluorescent Amplex® UltraRed reagent (A36006), which is oxidized by the H2O2-generated redox intermediates MPO-I and MPO-II to form a fluorescent product. The EnzChek® Myeloperoxidase Activity Assay Kit can be used to continuously detect these activities at room temperature over a broad dynamic range (1.5 to 200 ng/mL). The speed (30 minutes), sensitivity, and mix-and-read convenience make this kit ideal for measuring MPO activities and for high-throughput screening for MPO-specific inhibitors.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: Myeloperoxidase (MPO) Assay
Product Line: EnzChek®
Product Size: 1 kit
Substrate Properties: Chemical Substrate
Target Enzyme: Myeloperoxidase
Technique: Fluorescence Intensity
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products