Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

ELF™ 97 Immunohistochemistry Kit

Catalog number:  E6600

Invitrogen™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ELF® 97 Immunohistochemistry Kit can be used to detection biotinylated probes including secondary antibodies. The kit also includes the ELF® 97 phosphatase substrate that upon hydrolysis produces a bright and photostable yellow-green fluorescent precipitate at the site of enzymatic activity. This fluorescent precipitate has several unique spectral characteristics, including an extremely large Stokes shift, 180 nm that makes it easily distinguishable from endogenous fluorescence.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
Detection Method: Fluorescent
For Use With (Equipment): Fluorescence Microscope
Product Line: ELF®
Product Size: 1 kit
Reactivity: All species
Substrate: ELF® 97
Substrate Type: AP (Alkaline Phosphatase) Substrate
Target Species: All species
Target Specificity: Not Target-Specific
Technique: Enzyme-Labeled Fluorescence
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols