Invitrogen™

Novex™ TBE-Urea Gels, 6%, 10 well

Catalog number:  EC6865BOX

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Novex® TBE-Urea Gels are denaturing polyacrylamide gels that resolve single-stranded DNA oligos or RNA into sharp, distinct bands. These gels are optimized for the analysis and purification of products ranging from 20-800 bases, making them an ideal choice for synthetic oligo analysis and purification, RNase Protection Assays (RPA), in vitro transcription studies, and Northern blot analysis. Novex® TBE-Urea gels are designed to run on the XCell SureLock™ Mini-Cell.

Formulation: Novex® TBE-Urea gels are made with high-purity reagents, including Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, APS, and 7M urea. Strict quality control ensures that gels and buffers are DNase- and RNase-free.

Recommended Buffers: For optimum performance, Novex® TBE-Urea sample buffers and Novex® TBE Running Buffer are strongly recommended for use with these gels. For analytical applications, Novex® TBE-Urea Sample Buffer is recommended. This buffer contains urea, the density agent Ficoll™, which yields sharper, straighter bands than conventional density agents, and the tracking dyes Bromophenol Blue and Xylene Cyanol.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Percentage: 6 %
Gel Type: TBE-Urea
Product Line: Novex™
Product Size: 1 box
Wells: 10 wells
Shipping Condition: Wet Ice

Contents & storage

Catalog numbers for gels refer to a single box. One box contains 10 gels. Novex® TBE-Urea Gels have a shelf life of between 4 and 8 weeks depending upon gel type. Store at +4°C.

Documents

Manuals & protocols

Recommended products