New Protein Electrophoresis and Western Blotting Support Center—Find technical tips and troubleshooting recommendations from our scientists.

XCell SureLock™ Mini-Cell

Catalog number: EI0001

Invitrogen™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The XCell SureLock® Mini-Cell allows you to run Novex® mini-gels quickly, easily, and leak-free without any clamps or grease. The XCell line of mini-cells has always distinguished itself by being simple, sturdy, and convenient. The tradition continues with the patented XCell SureLock® Mini-Cell. The incorporation of a gel tension wedge, in place of the rear wedge used on earlier models, makes the XCell SureLock® Mini-Cell the easiest-to-use mini-cell on the market.

How the system works

The XCell SureLock® Mini-Cell holds gels firmly with a simple gel tension wedge. When the lever on the gel tension wedge is pushed forward into the locked position, an even, horizontal force is generated. This seals the gel/buffer core assembly firmly into position in the lower buffer chamber (see figure). The positive locking action of the gel tension wedge ensures a trouble-free, leak-free gel run every time.

Easy set-up procedure

The XCell SureLock® Mini-Cell can be easily set up in just three simple steps (see figure). The XCell SureLock® Mini-Cell is available separately or as a part of a complete package to fit your electrophoresis needs (see table).

Upgrade your XCell II™ to an XCell SureLock® Mini-Cell

Conversion of an XCell II™ into an XCell SureLock® is easy with the SureLock® Retrofit Kit (available separately). The kit consists of a SureLock® mini-cell lid, gel tension wedge, and molded mini-gel buffer dam.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): XCell SureLock® Mini
Capacity: Up to 2 mini-gels
Gel Compatibility: Novex® Mini Gels, NuPAGE® Gels
Gel Size: Mini (8 cm x 8 cm)
Product Line: XCell SureLock®
Product Size: 1 unit
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature. Warrantied for one year from the time of purchasing.

Documents

Manuals & protocols

Recommended products