XCell II™ Blot Module

Catalog number: EI9051

Novex™  Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The XCell II™ Blot Module allows you to easily transfer proteins or nucleic acids from mini-gels to membranes. It fits neatly into the XCell SureLock® and XCell II™ Mini-Cells in place of the gel/buffer core assembly. It requires less than 200 mL of transfer buffer for Western, Southern, and northern transfers. Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders. Maximum blot size is 9 cm x 9 cm.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Mode of Transfer: Wet
Gel Compatibility: NuPAGE® Gels, Bolt® Bis-Tris Plus Gels, Novex® Mini Gels
Running Dimension: Vertical
For Use With (Equipment): XCell SureLock® Mini
Capacity: Up to 2 mini-gels
Gel Size: Mini (8 cm x 8 cm)
Product Size: 1 unit
Shipping Condition: Room Temperature

Contents & storage

The XCell II™ Blot Module includes anode core, cathode core, sponge pads (8/pack), and instruction manual.

Documents

Manuals & protocols

Recommended products