Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

FilmTracer™ FM® 1-43 Green Biofilm Cell Stain

Catalog number: F10317

Molecular Probes™  Related applications: Microbiological Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

FM dyes are lipophilic styryl compounds used in a wide variety of studies involving plasma membrane and vesiculation. The water-soluble FM® dyes are virtually nonfluorescent in aqueous media and are believed to insert into the surface membrane where they become intensely fluorescent. FM® 1-43 stain has been used successfully to stain the cell bodies specifically in a complex biofilm mileu, including Pseudomonas aeruginosa, Escherichia coli, Staphylococcus sp., Acidothiobacillus caldus, and Vibrio cholerae.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Solid
Color: Green
Label or Dye: FM 1-43⁄lipid
Product Size: 1 mg
Emission Class: Visible
Detection Method: Fluorescent
Excitation Class: Visible
For Use With (Equipment): Microplate Reader, Fluorescence Microscope, Fluorescent Imager, Confocal Microscope, Flow Cytometer
Sub-Cellular Localization: Cell Membranes & Lipids
Flow Cytometer Laser Lines: 488
Excitation⁄Emission (nm): 472⁄580
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature; Dessicate; protect from light

Documents

Manuals & protocols

Recommended products