Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.
Invitrogen™

Fibrinogen From Human Plasma, Alexa Fluor™ 594 Conjugate

Catalog number:  F13193

 Related applications: Cell Viability, Proliferation & Function | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Molecular Probes® fibrinogen conjugates are prepared by attaching fluorescent dye molecules to purified human fibrinogen (approximately 15 dye molecules for each fibrinogen molecule), purifying the conjugate to remove unreacted dye, and then lyophilizing for storage.

Fluorescently labeled fibrinogen has proven to be a valuable tool for investigating platelet activation and subsequent fibrinogen binding. For instance, fluorescein-labeled fibrinogen has been used to detect fibrinogen bound to activated platelets by flow cytometry.

Human Fibrinogen Conjugate Specifications:
• Label (Ex/Em): Alexa Fluor® 594 (~590/617 nm)
• Spectrally similar to Texas Red® dye conjugates, but brighter
• Lyophilized product can be dissolved in buffer (e.g., sodium bicarbonate, pH 8.3) for use
• Fluorescence is typically detected using fluorescence microscopy or flow cytometry


Find More Probes for Cell Adhesion and Receptor Binding
Review Probes for Cell Adhesion, Chemotaxis, Multidrug Resistance and Glutathione—Section 15.6 and Probes for Following Receptor Binding and Phagocytosis—Section 16.1 in the Molecular Probes® Handbook for more information on these probes.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Alexa Fluor™
Product Size: 5 mg
Protein Subtype: Fibrinogen
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products