Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Fura-4F, AM, cell permeant - Special Packaging

Catalog number: F14175

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Fura-4F AM is an intracellular calcium indicator similar to fura-2 but with a response range shifted to detect higher Ca2+ concentrations. This acetoxymethyl (AM) ester derivative is useful for noninvasive intracellular loading. This sku can be used in place of the recently discontinued fura 5F AM, F14177. Ex/Ems are identical, and more importantly the Kd's(Ca2+) µM are very similar: 0.40 for fura-5F AM vs 0.77 for fura-4F AM.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Calcium Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope
Cell Permeability: Cell-Permeant
Detection Method: Fluorescent
Product Size: 10 x 50 µg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products