Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

FocalCheck™ Microspheres, 6 µm, fluorescent dark-red ring stain/green throughout

Catalog number: F14807

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The sharp ring stains exhibited by the 6.0 µm FocalCheck™ fluorescent dark-red ring stained/green throughout microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the ring image and disk image of the combination beads are perfectly coincident in all dimensions.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Calibration Type: Confocal Microscope Calibration
Color: Dark Red, Green
Diameter (Metric): 6 µm
Format: Suspension Beads
Product Line: FocalCheck™
Product Size: 0.5 mL
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols